Assay Details
| Gene Symbol: LOC4337945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782008.2 | XP_015637494.1 | 3 - 4 | 1238 | 86 | |
| XM_015782009.2 | XP_015637495.1 | 3 - 4 | 1144 | 86 | ||
| GenBank mRNA | AK103471.1 | - | 3 - 4 | 1142 | 86 | |
More Information
Set Membership: |
|

LOC4337945
Genomic Map