Assay Details
| Gene Symbol: LOC4338348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784704.2 | XP_015640190.1 | 7 - 8 | 1344 | 114 | |
| GenBank mRNA | AK070329.1 | - | 7 - 8 | 1335 | 114 | |
More Information
Set Membership: |
|

LOC4338348
Genomic Map