Assay Details
| Gene Symbol: LOC4338348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784704.2 | XP_015640190.1 | 8 - 9 | 1503 | 113 | |
| GenBank mRNA | AK059501.1 | - | 2 - 3 | 218 | 113 | |
| AK070329.1 | - | 8 - 9 | 1494 | 113 | ||
| AK104884.1 | - | 2 - 3 | 218 | 113 | ||
More Information
Set Membership: |
|

LOC4338348
Genomic Map