Assay Details
| Gene Symbol: LOC4338348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784704.2 | XP_015640190.1 | 9 - 10 | 1730 | 83 | |
| GenBank mRNA | AK059501.1 | - | 3 - 4 | 445 | 83 | |
| AK070329.1 | - | 9 - 10 | 1721 | 83 | ||
| AK104884.1 | - | 3 - 4 | 445 | 83 | ||
More Information
Set Membership: |
|

LOC4338348
Genomic Map