Assay Details
| Gene Symbol: LOC4339038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782748.2 | XP_015638234.1 | 3 - 4 | 563 | 82 | |
| GenBank mRNA | AK108033.1 | - | 3 - 4 | 569 | 82 | |
| Gene Symbol: LOC4339132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK073132.1 | - | 655 | 82 | ||
More Information
Set Membership: |
|

LOC4339038
Genomic Map