Assay Details
| Gene Symbol: LOC4340655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788779.2 | XP_015644265.1 | 1 - 2 | 209 | 112 | |
| XM_015788780.2 | XP_015644266.1 | 1 - 2 | 63 | 112 | ||
| GenBank mRNA | AK240679.1 | - | 1 - 2 | 178 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788780.2 transcripts.
Set Membership: |
|

LOC4340655
Genomic Map