Assay Details
| Gene Symbol: LOC4340655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788779.2 | XP_015644265.1 | 5 - 6 | 720 | 147 | |
| XM_015788780.2 | XP_015644266.1 | 5 - 6 | 515 | 147 | ||
| GenBank mRNA | AK240679.1 | - | 5 - 6 | 728 | 147 | |
More Information
Set Membership: |
|

LOC4340655
Genomic Map