Assay Details
| Gene Symbol: LOC4341663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786513.2 | XP_015641999.1 | 10 - 11 | 1154 | 94 | |
| GenBank mRNA | AK063383.1 | - | 10 - 11 | 1030 | 94 | |
| AY222862.1 | - | 10 - 11 | 904 | 94 | ||
More Information
Set Membership: |
|

LOC4341663
Genomic Map