Assay Details
| Gene Symbol: LOC4341663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786513.2 | XP_015641999.1 | 11 - 12 | 1230 | 101 | |
| GenBank mRNA | AK063383.1 | - | 11 - 12 | 1106 | 101 | |
| AY222862.1 | - | 11 - 12 | 980 | 101 | ||
More Information
Set Membership: |
|

LOC4341663
Genomic Map