Assay Details
| Gene Symbol: LOC4341709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785854.2 | XP_015641340.1 | 9 - 10 | 2479 | 100 | |
| GenBank mRNA | KF999959.1 | - | 9 - 10 | 2479 | 100 | |
More Information
Set Membership: |
|

LOC4341709
Genomic Map