Assay Details
| Gene Symbol: LOC4343738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789260.2 | XP_015644746.1 | 1 - 2 | 687 | 91 | |
| GenBank mRNA | AK072014.1 | - | 1 - 2 | 674 | 91 | |
| AK072176.1 | - | 1 - 2 | 675 | 91 | ||
| AK105268.1 | - | 1 - 2 | 674 | 91 | ||
More Information
Set Membership: |
|

LOC4343738
Genomic Map