Assay Details
| Gene Symbol: LOC4343946 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790892.1 | XP_015646378.1 | 4 - 5 | 1553 | 78 | |
| GenBank mRNA | AB122058.1 | - | 4 - 5 | 1417 | 78 | |
| AK069968.1 | - | 4 - 5 | 1405 | 78 | ||
| AK105687.1 | - | 4 - 5 | 1404 | 78 | ||
More Information
Set Membership: |
|

LOC4343946
Genomic Map