Assay Details
| Gene Symbol: LOC4344397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791070.2 | XP_015646556.1 | 4 - 5 | 532 | 90 | |
| GenBank mRNA | AB050724.1 | - | 4 - 5 | 470 | 90 | |
| AB053297.1 | - | 4 - 5 | 475 | 90 | ||
| AK061715.1 | - | 4 - 5 | 488 | 90 | ||
| AK068430.1 | - | 4 - 5 | 484 | 90 | ||
| AK102751.1 | - | 488 | 90 | |||
| GQ848056.1 | - | 4 - 5 | 469 | 90 | ||
More Information
Set Membership: |
|

LOC4344397
Genomic Map