Assay Details
| Gene Symbol: LOC4344397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791070.2 | XP_015646556.1 | 6 - 7 | 700 | 84 | |
| GenBank mRNA | AB050724.1 | - | 6 - 7 | 638 | 84 | |
| AB053297.1 | - | 6 - 7 | 643 | 84 | ||
| AK061715.1 | - | 6 - 7 | 656 | 84 | ||
| AK102751.1 | - | 656 | 84 | |||
| GQ848056.1 | - | 6 - 7 | 637 | 84 | ||
More Information
Set Membership: |
|

LOC4344397
Genomic Map