Assay Details
| Gene Symbol: LOC4347311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756309.2 | XP_015611795.1 | 10 - 11 | 1570 | 97 | |
| GenBank mRNA | AB107218.1 | - | 10 - 11 | 1495 | 97 | |
| AK068061.1 | - | 10 - 11 | 1571 | 97 | ||
More Information
Set Membership: |
|

LOC4347311
Genomic Map