Assay Details
| Gene Symbol: LOC4347311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756309.2 | XP_015611795.1 | 12 - 13 | 1813 | 55 | |
| GenBank mRNA | AB107218.1 | - | 12 - 13 | 1738 | 55 | |
| AK068061.1 | - | 12 - 13 | 1814 | 55 | ||
More Information
Set Membership: |
|

LOC4347311
Genomic Map