Assay Details
| Gene Symbol: LOC4347311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756309.2 | XP_015611795.1 | 13 - 14 | 1873 | 72 | |
| GenBank mRNA | AB107218.1 | - | 13 - 14 | 1798 | 72 | |
| AK068061.1 | - | 13 - 14 | 1874 | 72 | ||
| BT131688.1 | - | 1 - 2 | 55 | 72 | ||
More Information
Set Membership: |
|

LOC4347311
Genomic Map