Assay Details
| Gene Symbol: LOC4346851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756376.2 | XP_015611862.1 | 2 - 3 | 267 | 65 | |
| GenBank mRNA | AK060652.1 | - | 2 - 3 | 216 | 65 | |
| AK061261.1 | - | 2 - 3 | 202 | 65 | ||
More Information
Set Membership: |
|

LOC4346851
Genomic Map