Assay Details
| Gene Symbol: LOC4347610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756807.2 | XP_015612293.1 | 10 - 11 | 1622 | 61 | |
| GenBank mRNA | AK060889.1 | - | 3 - 4 | 294 | 61 | |
| AK101403.1 | - | 10 - 11 | 1614 | 61 | ||
More Information
Set Membership: |
|

LOC4347610
Genomic Map