Assay Details
| Gene Symbol: LOC4347610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756807.2 | XP_015612293.1 | 5 - 6 | 1020 | 88 | |
| GenBank mRNA | AK101403.1 | - | 5 - 6 | 1012 | 88 | |
More Information
Set Membership: |
|

LOC4347610
Genomic Map