Assay Details
| Gene Symbol: LOC4347745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756659.1 | XP_015612145.1 | 2 - 3 | 1382 | 82 | |
| GenBank mRNA | AK100579.1 | - | 2 - 3 | 1308 | 82 | |
More Information
Set Membership: |
|

LOC4347745
Genomic Map