Assay Details
| Gene Symbol: LOC4347814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756210.2 | XP_015611696.1 | 11 - 12 | 1447 | 109 | |
| XM_015756211.2 | XP_015611697.1 | 10 - 11 | 1333 | 109 | ||
| GenBank mRNA | AB159444.1 | - | 11 - 12 | 1442 | 109 | |
| AK101142.1 | - | 11 - 12 | 1442 | 109 | ||
More Information
Set Membership: |
|

LOC4347814
Genomic Map