Assay Details
| Gene Symbol: LOC4347814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756210.2 | XP_015611696.1 | 5 - 6 | 616 | 106 | |
| GenBank mRNA | AB159444.1 | - | 5 - 6 | 611 | 106 | |
| AK101142.1 | - | 5 - 6 | 611 | 106 | ||
More Information
Set Membership: |
|

LOC4347814
Genomic Map