Assay Details
| Gene Symbol: LOC4348803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757515.2 | XP_015613001.1 | 5 - 6 | 399 | 133 | |
| GenBank mRNA | AK059744.1 | - | 5 - 6 | 397 | 133 | |
| EU152202.1 | - | 5 - 6 | 374 | 154 | ||
More Information
Set Membership: |
|

LOC4348803
Genomic Map