Assay Details
| Gene Symbol: LOC4349053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759345.2 | XP_015614831.1 | 7 - 8 | 1566 | 103 | |
| GenBank mRNA | AK073085.1 | - | 7 - 8 | 1564 | 103 | |
| AK101354.1 | - | 4 - 5 | 583 | 103 | ||
More Information
Set Membership: |
|

LOC4349053
Genomic Map