Assay Details
| Gene Symbol: LOC4352866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762955.2 | XP_015618441.1 | 12 - 13 | 1436 | 103 | |
| GenBank mRNA | AK063915.1 | - | 12 - 13 | 1438 | 103 | |
| AK067318.1 | - | 12 - 13 | 1642 | 103 | ||
| AK103431.1 | - | 12 - 13 | 1406 | 103 | ||
More Information
Set Membership: |
|

LOC4352866
Genomic Map