Assay Details
| Gene Symbol: LOC4352866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762955.2 | XP_015618441.1 | 4 - 5 | 794 | 78 | |
| GenBank mRNA | AK063915.1 | - | 4 - 5 | 796 | 78 | |
| AK067318.1 | - | 4 - 5 | 800 | 78 | ||
| AK103431.1 | - | 4 - 5 | 764 | 78 | ||
More Information
Set Membership: |
|

LOC4352866
Genomic Map