Assay Details
| Gene Symbol: LOC4352866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762955.2 | XP_015618441.1 | 5 - 6 | 849 | 73 | |
| GenBank mRNA | AK063915.1 | - | 5 - 6 | 851 | 73 | |
| AK067318.1 | - | 5 - 6 | 855 | 73 | ||
| AK103431.1 | - | 5 - 6 | 819 | 73 | ||
More Information
Set Membership: |
|

LOC4352866
Genomic Map