Assay Details
| Gene Symbol: LOC4326806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767029.2 | XP_015622515.1 | 5 - 6 | 651 | 113 | |
| GenBank mRNA | AK066111.1 | - | 5 - 6 | 658 | 113 | |
More Information
Set Membership: |
|

LOC4326806
Genomic Map