Assay Details
| Gene Symbol: LOC4325046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766956.2 | XP_015622442.1 | 13 - 14 | 1977 | 97 | |
| GenBank mRNA | AK102226.1 | - | 13 - 14 | 1954 | 97 | |
More Information
Set Membership: |
|

LOC4325046
Genomic Map