Assay Details
| Gene Symbol: LOC4325046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766956.2 | XP_015622442.1 | 9 - 10 | 1595 | 101 | |
| GenBank mRNA | AK102226.1 | - | 9 - 10 | 1572 | 101 | |
More Information
Set Membership: |
|

LOC4325046
Genomic Map