Assay Details
| Gene Symbol: LOC4324927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766839.2 | XP_015622325.1 | 5 - 6 | 1171 | 88 | |
| GenBank mRNA | AB731703.1 | - | 5 - 6 | 953 | 88 | |
| AK073453.1 | - | 5 - 6 | 986 | 88 | ||
More Information
Set Membership: |
|

LOC4324927
Genomic Map