Assay Details
| Gene Symbol: LOC4327247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB003327.1 | - | 3 - 4 | 399 | 99 | |
| AK066160.1 | - | 3 - 4 | 452 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4327247| Gene Symbol: LOC4327247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB003327.1 | - | 3 - 4 | 399 | 99 | |
| AK066160.1 | - | 3 - 4 | 452 | 99 | ||
Set Membership: |
|