Assay Details
| Gene Symbol: LOC4332694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772345.2 | XP_015627831.1 | 3 - 4 | 528 | 82 | |
| GenBank mRNA | AK105342.1 | - | 3 - 4 | 453 | 82 | |
More Information
Set Membership: |
|

LOC4332694
Genomic Map