Assay Details
| Gene Symbol: LOC4333020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775047.2 | XP_015630533.1 | 5 - 6 | 637 | 73 | |
| GenBank mRNA | AB110190.1 | - | 5 - 6 | 592 | 73 | |
| AK103600.1 | - | 5 - 6 | 594 | 73 | ||
More Information
Set Membership: |
|

LOC4333020
Genomic Map