Assay Details
| Gene Symbol: LOC4333841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775411.1 | XP_015630897.1 | 4 - 5 | 657 | 104 | |
| GenBank mRNA | AK100165.1 | - | 4 - 5 | 544 | 104 | |
| AK105026.2 | - | 4 - 5 | 541 | 104 | ||
| KF214527.1 | - | 4 - 5 | 534 | 104 | ||
| KM982771.1 | - | 4 - 5 | 516 | 104 | ||
| U28047.3 | - | 4 - 5 | 549 | 104 | ||
More Information
Set Membership: |
|

LOC4333841
Genomic Map