Assay Details
| Gene Symbol: LOC4333841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775411.1 | XP_015630897.1 | 6 - 7 | 986 | 125 | |
| GenBank mRNA | AK059920.1 | - | 2 - 3 | 314 | 125 | |
| AK100165.1 | - | 6 - 7 | 873 | 125 | ||
| AK103027.1 | - | 6 - 7 | 869 | 125 | ||
| AK105026.2 | - | 6 - 7 | 870 | 125 | ||
| KF214527.1 | - | 6 - 7 | 863 | 125 | ||
| KM982771.1 | - | 6 - 7 | 845 | 125 | ||
| U28047.3 | - | 6 - 7 | 878 | 125 | ||
More Information
Set Membership: |
|

LOC4333841
Genomic Map