Assay Details
| Gene Symbol: LOC4334112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772510.2 | XP_015627996.1 | 3 - 4 | 441 | 68 | |
| GenBank mRNA | AK061165.1 | - | 3 - 4 | 429 | 68 | |
More Information
Set Membership: |
|

LOC4334112
Genomic Map