Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 11 - 12 | 1806 | 99 | |
| XM_015770432.2 | XP_015625918.1 | 12 - 13 | 1725 | 99 | ||
| GenBank mRNA | AK066478.1 | - | 2 - 3 | 225 | 99 | |
| AK067099.1 | - | 11 - 12 | 1770 | 99 | ||
| AK103515.1 | - | 11 - 12 | 1502 | 99 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map