Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 15 - 16 | 2239 | 80 | |
| XM_015770432.2 | XP_015625918.1 | 16 - 17 | 2158 | 80 | ||
| GenBank mRNA | AK066478.1 | - | 6 - 7 | 658 | 80 | |
| AK067099.1 | - | 15 - 16 | 2203 | 80 | ||
| AK103515.1 | - | 15 - 16 | 1935 | 80 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map