Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 16 - 17 | 2386 | 106 | |
| XM_015770432.2 | XP_015625918.1 | 17 - 18 | 2305 | 106 | ||
| GenBank mRNA | AK066478.1 | - | 7 - 8 | 805 | 106 | |
| AK067099.1 | - | 16 - 17 | 2350 | 106 | ||
| AK103515.1 | - | 16 - 17 | 2082 | 106 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map