Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 20 - 21 | 2985 | 65 | |
| XM_015770432.2 | XP_015625918.1 | 21 - 22 | 2904 | 65 | ||
| GenBank mRNA | AK066478.1 | - | 11 - 12 | 1404 | 65 | |
| AK067099.1 | - | 20 - 21 | 2949 | 65 | ||
| AK103515.1 | - | 20 - 21 | 2681 | 65 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map