Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 22 - 23 | 3203 | 142 | |
| XM_015770432.2 | XP_015625918.1 | 23 - 24 | 3122 | 142 | ||
| GenBank mRNA | AK066478.1 | - | 13 - 14 | 1622 | 142 | |
| AK067099.1 | - | 22 - 23 | 3167 | 142 | ||
| AK103515.1 | - | 22 - 23 | 2899 | 142 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map