Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 23 - 24 | 3289 | 141 | |
| XM_015770432.2 | XP_015625918.1 | 24 - 25 | 3208 | 141 | ||
| GenBank mRNA | AK066478.1 | - | 14 - 15 | 1708 | 141 | |
| AK067099.1 | - | 23 - 24 | 3253 | 141 | ||
| AK103515.1 | - | 23 - 24 | 2985 | 141 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map