Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 28 - 29 | 3700 | 129 | |
| XM_015770432.2 | XP_015625918.1 | 29 - 30 | 3619 | 129 | ||
| GenBank mRNA | AK066478.1 | - | 18 - 19 | 2326 | 129 | |
| AK067099.1 | - | 28 - 29 | 3664 | 129 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map