Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 30 - 31 | 3883 | 131 | |
| XM_015770432.2 | XP_015625918.1 | 31 - 32 | 3802 | 131 | ||
| GenBank mRNA | AK066478.1 | - | 20 - 21 | 2509 | 131 | |
| AK067099.1 | - | 30 - 31 | 3847 | 131 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map