Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 9 - 10 | 1528 | 68 | |
| XM_015770432.2 | XP_015625918.1 | 10 - 11 | 1447 | 68 | ||
| GenBank mRNA | AK067099.1 | - | 9 - 10 | 1492 | 68 | |
| AK103515.1 | - | 9 - 10 | 1224 | 68 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map