Assay Details
| Gene Symbol: LOC4330285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769956.2 | XP_015625442.1 | 3 - 4 | 610 | 60 | |
| GenBank mRNA | AK059611.1 | - | 3 - 4 | 579 | 60 | |
| AK099179.1 | - | 3 - 4 | 577 | 60 | ||
More Information
Set Membership: |
|

LOC4330285
Genomic Map