Assay Details
| Gene Symbol: LOC4332006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777784.2 | XP_015633270.1 | 1 - 2 | 337 | 64 | |
| GenBank mRNA | AK111713.1 | - | 1 - 2 | 338 | 64 | |
| AK111888.1 | - | 1 - 2 | 220 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777784.2 transcripts.
Set Membership: |
|

LOC4332006
Genomic Map