Assay Details
| Gene Symbol: LOC107277011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776333.2 | XP_015631819.1 | 18 - 19 | 3182 | 108 | |
| GenBank mRNA | AK288069.1 | - | 6 - 7 | 822 | 108 | |
More Information
Set Membership: |
|

LOC107277011
Genomic Map